Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NPNT Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551988
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551988 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551988 Supplier Invitrogen™ Supplier No. PA551988
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RQCVNTFGSY ICKCHKGFDL MYIGGKYQCH DIDECSLGQY QCSSFARCYN VRGSYKCKCK EGYQGDGLTC VYIPKVMIEP SGPIHVPKGN GTILKGDTGN NNWIPDVGST WWPPKTPYIP PIITNRPTSK PTTRPTPK Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 54%.

Functional ligand of integrin alpha-8/beta-1 in kidney development. Regulates the expression of GDNF with integrin alpha-8/beta-1 which is essential for kidney development. May also play a role in the development and function of various tissues, regulating cell adhesion, spreading and survival through the binding of several integrins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NPNT
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NPNT
Gene Accession No. Q6UXI9
Gene Alias 1110009H02Rik; AA682063; AI314031; EGFL6L; Nctn; Neph1; Nephronectin; NPNT; POEM; Preosteoblast EGF-like repeat protein with MAM domain; Protein EGFL6-like; UNQ295/PRO334
Gene Symbols NPNT
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NPNT. Recombinant protein control fragment (Product #RP-101074).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 255743
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.