Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NPNT Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA565600

Catalog No. PIPA565600


Only null left
Add to cart

Description

Description

Immunogen sequence: TVDNRVQTDPQ KPRGDVFIPR QPSNDLFEIF EIERGVSADD EAKDDPGVLV HSCNFDHGLC GWIREKDNDL HWEPIRDPAG GQYLTVSAAK APGGKAARLV LPLGRLMHSG DLCLSFRHKV TGLHSGTLQV FVRKHGAHGA ALWGRNGGHG WRQTQITL Highest antigen sequence identity to the following orthologs - mouse 92%, rat 94%.

Functional ligand of integrin alpha-8/beta-1 in kidney development. Regulates the expression of GDNF with integrin alpha-8/beta-1 which is essential for kidney development. May also play a role in the development and function of various tissues, regulating cell adhesion, spreading and survival through the binding of several integrins.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

NPNT
Polyclonal
Unconjugated
NPNT
1110009H02Rik; AA682063; AI314031; EGFL6L; Nctn; Neph1; Nephronectin; NPNT; POEM; Preosteoblast EGF-like repeat protein with MAM domain; Protein EGFL6-like; UNQ295/PRO334
Rabbit
Antigen affinity chromatography
RUO
255743
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot, Immunocytochemistry
0.10 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q6UXI9
NPNT
Recombinant Human NPNT. Recombinant protein control fragment (Product #RP-104893).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.