Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NPNT Polyclonal Antibody
Goat Polyclonal Antibody
Supplier: Invitrogen™ PA547610
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
Functional ligand of integrin alpha-8/beta-1 in kidney development. Regulates the expression of GDNF with integrin alpha-8/beta-1 which is essential for kidney development. May also play a role in the development and function of various tissues, regulating cell adhesion, spreading and survival through the binding of several integrins.
Specifications
NPNT | |
Polyclonal | |
Unconjugated | |
NPNT | |
1110009H02Rik; AA682063; AI314031; EGFL6L; Nctn; Neph1; Nephronectin; NPNT; POEM; Preosteoblast EGF-like repeat protein with MAM domain; Protein EGFL6-like; UNQ295/PRO334 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
114249 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q91V88 | |
NPNT | |
Mouse myeloma cell line NS0-derived recombinant mouse Nephronectin Asp20-Cys561. | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction