Learn More
NPPA Mouse anti-Human, Clone: OTI4H4, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF809559

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The protein encoded by this gene belongs to the natriuretic peptide family. Natriuretic peptides are implicated in the control of extracellular fluid volume and electrolyte homeostasis. This protein is synthesized as a large precursor (containing a signal peptide), which is processed to release a peptide from the N-terminus with similarity to vasoactive peptide, cardiodilatin, and another peptide from the C-terminus with natriuretic-diuretic activity. Mutations in this gene have been associated with atrial fibrillation familial type 6.Specifications
NPPA | |
Monoclonal | |
Unconjugated | |
NPPA | |
ANF, ANP, ATFB6, ATRST2, CDD, CDD-ANF, CDP, PND | |
Mouse | |
Affinity Chromatography | |
RUO | |
4878 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
OTI4H4 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P01160 | |
NPPA | |
Human recombit protein fragment corresponding to amino acids 26-153 of human NPPA produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.