Learn More
Invitrogen™ NPR1 Polyclonal Antibody

Description
The antibody detects endogenous levels of total NPR1 protein.
Specifications
Specifications
| Antigen | NPR1 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 1.3 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene | NPR1 |
| Gene Accession No. | P16066, P18293, P18910 |
| Gene Alias | AI893888; ANPa; ANP-A; ANPRA; ANPR-A; atrial natriuretic peptide A-type receptor; atrial natriuretic peptide receptor 1; atrial natriuretic peptide receptor A; atrial natriuretic peptide receptor type A; atrionatriuretic peptide receptor A; Gca; GC-A; Guanylate cyclase A; guanylyl cyclase-A; GUC2A; GUCY2A; natriuretic peptide A type receptor; natriuretic peptide receptor 1; natriuretic peptide receptor A; natriuretic peptide receptor A/guanylate cyclase A (atrionatriuretic peptide receptor A); Nppa receptor; Npr1; Npra; NPR-A; Pndr; testicular tissue protein Li 20 |
| Gene Symbols | NPR1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.