Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Npr2 Recombinant Rabbit Monoclonal Antibody (19C7)
GREENER_CHOICE

Catalog No. FEMA550136
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550136 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA550136 Supplier Invitrogen™ Supplier No. MA550136
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Npr2 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow, ELISA

This gene encodes natriuretic peptide receptor B, one of two integral membrane receptors for natriuretic peptides. Both NPR1 and NPR2 contain five functional domains: an extracellular ligand-binding domain, a single membrane-spanning region, and intracellularly a protein kinase homology domain, a helical hinge region involved in oligomerization, and a carboxyl-terminal guanylyl cyclase catalytic domain. The protein is the primary receptor for C-type natriuretic peptide, which upon ligand binding exhibits greatly increased guanylyl cyclase activity. Mutations in this gene are the cause of acromesomelic dysplasia Maroteaux type.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Npr2
Applications ELISA, Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 19C7
Concentration 0.8 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.05% BSA, 150mM NaCl and 0.02% sodium azide; pH 7.4
Gene Npr2
Gene Accession No. P20594
Gene Alias AMDM; ANPb; ANP-B; ANPRB; ANPR-B; atrial natriuretic peptide B-type receptor; atrial natriuretic peptide receptor 2; atrial natriuretic peptide receptor B; atrial natriuretic peptide receptor type B; cn; ECDM; GC-B; GC-B2; GC-B3; guanylate cyclase 2B; guanylate cyclase B; guanylyl cyclase-B; GUC2B; GUCY2B; mNPR-B; natriuretic peptide receptor 2; natriuretic peptide receptor B type; natriuretic peptide receptor B/guanylate cyclase B (atrionatriuretic peptide receptor B); natriuretic peptide receptor-B; Nppb receptor; NPR2; NPRB; NPR-B; NPRBi; pwe; SNSK
Gene Symbols Npr2
Host Species Rabbit
Immunogen A synthesized peptide derived from Human NPR2.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4882
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.