Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Npr2 Recombinant Rabbit Monoclonal Antibody (1S1H1)
GREENER_CHOICE

Catalog No. PIMA537887
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA537887 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA537887 Supplier Invitrogen™ Supplier No. MA537887
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Positive Samples: Mouse lung, Mouse spleen Immunogen sequence: NLLLRMEQYA NNLEKLVEER TQAYLEEKRK AEALLYQILP HSVAEQLKRG ETVQAEAFDS VTIYFSDIVG FTALSAESTP MQVVTLLNDL YTCFDAIIDN F.

This gene encodes natriuretic peptide receptor B, one of two integral membrane receptors for natriuretic peptides. Both NPR1 and NPR2 contain five functional domains: an extracellular ligand-binding domain, a single membrane-spanning region, and intracellularly a protein kinase homology domain, a helical hinge region involved in oligomerization, and a carboxyl-terminal guanylyl cyclase catalytic domain. The protein is the primary receptor for C-type natriuretic peptide, which upon ligand binding exhibits greatly increased guanylyl cyclase activity. Mutations in this gene are the cause of acromesomelic dysplasia Maroteaux type.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Npr2
Applications ELISA, Western Blot
Classification Recombinant Monoclonal
Clone 1S1H1
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Npr2
Gene Accession No. Q6VVW5
Gene Alias AMDM; ANPb; ANP-B; ANPRB; ANPR-B; atrial natriuretic peptide B-type receptor; atrial natriuretic peptide receptor 2; atrial natriuretic peptide receptor B; atrial natriuretic peptide receptor type B; cn; ECDM; GC-B; GC-B2; GC-B3; guanylate cyclase 2B; guanylate cyclase B; guanylyl cyclase-B; GUC2B; GUCY2B; mNPR-B; natriuretic peptide receptor 2; natriuretic peptide receptor B type; natriuretic peptide receptor B/guanylate cyclase B (atrionatriuretic peptide receptor B); natriuretic peptide receptor-B; Nppb receptor; NPR2; NPRB; NPR-B; NPRBi; pwe; SNSK
Gene Symbols Npr2
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 800-900 of human ANPRB/NRP2 (P20594).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 230103
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.