Learn More
NPR3 Monoclonal Antibody (OTI4H1), TrueMAB™, OriGene
SDP
Mouse Monoclonal Antibody
Supplier:  OriGene TA501044
Encompass_Preferred
 View more versions of this product
Catalog No. 50-167-2187
Only null left
Add to Cart
Description
Description
ANPC is a receptor for atrial natriuretic peptide. It does not exhibit guanylate cyclase activity. There seem to be at least three ANP receptors: two with guanylate cyclase activity (ANPA and ANPB) and one (ANPC) which is probably responsible for the clearance of ANP from the circulation without a role in signal transduction.
Specifications
Specifications
NPR3
Monoclonal
Unconjugated
NPR3
ANP-C, ANPR-C, ANPRC, C5orf23, GUCY2B, NPR-C, NPRC
Mouse
Affinity Chromatography
RUO
4883
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
OTI4H1
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P17342
NPR3
Full length human recombit protein of human NPR3 produced in HEK293T cell.
100 μL
Primary
Human
Antibody
IgG2b
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.