Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NPR3 Mouse anti-Human, Clone: OTI11H6, liquid, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene TA501080S

Description
ANPC is a receptor for atrial natriuretic peptide. It does not exhibit guanylate cyclase activity. There seem to be at least three ANP receptors: two with guanylate cyclase activity (ANPA and ANPB) and one (ANPC) which is probably responsible for the clearance of ANP from the circulation without a role in signal transduction.Specifications
NPR3 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
P17342 | |
NPR3 | |
Full length human recombit protein of human NPR3 produced in HEK293T cell. | |
30 μL | |
Primary | |
Human | |
Antibody | |
IgG2b |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
OTI11H6 | |
Unconjugated | |
NPR3 | |
ANP-C, ANPR-C, ANPRC, C5orf23, GUCY2B, NPR-C, NPRC | |
Mouse | |
Affinity Chromatography | |
RUO | |
4883 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction