Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NPR3 Mouse anti-Human, Clone: OTI2B12, lyophilized, TrueMAB™
SDP

Catalog No. 501660083
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-166-0083 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-166-0083 Supplier OriGene Supplier No. CF501045
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO

ANPC is a receptor for atrial natriuretic peptide. It does not exhibit guanylate cyclase activity. There seem to be at least three ANP receptors: two with guanylate cyclase activity (ANPA and ANPB) and one (ANPC) which is probably responsible for the clearance of ANP from the circulation without a role in signal transduction.

Specifications

Antigen NPR3
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
Classification Monoclonal
Clone OTI2B12
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene NPR3
Gene Accession No. P17342
Gene Alias ANP-C, ANPR-C, ANPRC, C5orf23, GUCY2B, NPR-C, NPRC
Gene Symbols NPR3
Host Species Mouse
Immunogen Full length human recombit protein of human NPR3 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4883
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.