Learn More
NPR3 Mouse anti-Human, Clone: OTI4H1, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF501044

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
ANPC is a receptor for atrial natriuretic peptide. It does not exhibit guanylate cyclase activity. There seem to be at least three ANP receptors: two with guanylate cyclase activity (ANPA and ANPB) and one (ANPC) which is probably responsible for the clearance of ANP from the circulation without a role in signal transduction.Specifications
NPR3 | |
Monoclonal | |
Unconjugated | |
NPR3 | |
ANP-C, ANPR-C, ANPRC, C5orf23, GUCY2B, NPR-C, NPRC | |
Mouse | |
Affinity Chromatography | |
RUO | |
4883 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
OTI4H1 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P17342 | |
NPR3 | |
Full length human recombit protein of human NPR3 produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.