Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NPRA/NPR1 Antibody (OTI7H7), DyLight 550, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273044R
Description
NPRA/NPR1 Monoclonal specifically detects NPRA/NPR1 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
NPRA/NPR1 | |
Monoclonal | |
DyLight 550 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
NPR1 | |
Recombinant protein fragment corresponding to amino acids 746-1006 of human NPRA/NPR1 (NP_000897) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Western Blot | |
OTI7H7 | |
Western Blot | |
ANPa, ANP-A, ANPR-A, ANPRAEC 4.6.1.2, atrial natriuretic peptide receptor 1, Atrial natriuretic peptide receptor type A, atrionatriuretic peptide receptor A, GC-A, Guanylate cyclase A, GUC2A, GUCY2A, natriuretic peptide A type receptor, natriuretic peptide receptor A, natriuretic peptide receptor A/guanylate cyclase A (atrionatriuretic peptidereceptor A), NPRA, NPR-A | |
Mouse | |
Affinity Purified | |
Protein Kinase | |
4881.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction