Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NPRB/NPR2 Antibody (SR1075), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP322358100UL
Description
NPRB/NPR2 Monoclonal antibody specifically detects NPRB/NPR2 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow CytometrySpecifications
NPRB/NPR2 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Flow Cytometry | |
SR1075 | |
Western Blot 1:500-1:2000, Flow Cytometry 1:50 | |
AMD1;AMDM;ANPb;ANPRB;Atrial natriuretic peptide receptor 2;ECDM;GCB;GC-B;GUC2B;GUCY2B;NPRB;NPRBi;SNSK | |
A synthesized peptide derived from human NPRB/NPR2 (Uniprot #: P20594) | |
100 μg | |
Cardiovascular Biology, GPCR, Neuroscience, Signal Transduction | |
4882 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction