Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NPRB/NPR2 Rabbit anti-Human, Mouse, Rat, Clone: 1S1H1, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP315718100UL
This item is not returnable.
View return policy
Description
NPRB/NPR2 Monoclonal antibody specifically detects NPRB/NPR2 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
NPRB/NPR2 | |
Monoclonal | |
Unconjugated | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
A synthetic peptide corresponding to a sequence within amino acids 800-900 of human NPRB/NPR2 (P20594). NLLLRMEQYANNLEKLVEERTQAYLEEKRKAEALLYQILPHSVAEQLKRGETVQAEAFDSVTIYFSDIVGFTALSAESTPMQVVTLLNDLYTCFDAIIDNF | |
100 μg | |
Cardiovascular Biology, GPCR, Neuroscience, Signal Transduction | |
4882 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Western Blot | |
1S1H1 | |
Western Blot 1:500 - 1:2000 | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction