Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NPRC/Natriuretic Peptide Receptor C Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501984642
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-4642 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-4642 Supplier Bioss Supplier No. BS2333R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene encodes one of three natriuretic peptide receptors. Natriutetic peptides are small peptides which regulate blood volume and pressure, pulmonary hypertension, and cardiac function as well as some metabolic and growth processes. The product of this gene encodes a natriuretic peptide receptor responsible for clearing circulating and extracellular natriuretic peptides through endocytosis of the receptor. Multiple transcript variants encoding different isoforms have been found for this gene.

Specifications

Antigen NPRC/Natriuretic Peptide Receptor C
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene NPR3
Gene Accession No. P17342
Gene Alias A NPR-C; ANP-C; ANP-CR; ANPRC; ANPR-C; Atrial natriuretic peptide clearance receptor; Atrial natriuretic peptide clearence receptor 3; atrial natriuretic peptide C-type receptor; Atrial natriuretic peptide receptor 3; Atrial natriuretic peptide receptor type C; atrionatriuretic peptide receptor C; C5orf23; EF-2; guanylate cyclase C; GUCY2B; lgj; longjohn; natriuretic peptide receptor 3; natriuretic peptide receptor C/guanylate cyclase C (atrionatriuretic peptide receptor C); Nppc receptor; Npr3; NPRC; Npr-c; stri; strigosus
Gene Symbols NPR3
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human NPR-C.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4883
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.