Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NPRL3 Polyclonal Antibody

Catalog No. PIPA552849
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552849 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552849 Supplier Invitrogen™ Supplier No. PA552849

Rabbit Polyclonal Antibody

Immunogen sequence: GDSRFSDVIL ATILATKSEM CGQKFELKID NVRFVGHPTL LQHALGQISK TDPSPKREAP TMILFNVVFA LRANADPSVI NCLHNLSRRI ATVLQHEERR CQYLTREAKL ILALQDEVSA MADGNEGPQS PFHHILPKCK Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 97%.

The function of the encoded protein is not known.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NPRL3
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NPRL3
Gene Accession No. Q12980
Gene Alias -14 gene protein; Aag; alpha-globin regulatory element-containing gene protein; alphaRE-14; C16orf35; CGTHBA; conserved gene telomeric to alpha globin cluster; GATOR complex protein NPRL3; HS-26; HS40; HS-40; MARE; nitrogen permease regulator 3-like protein; NPR3; NPR3 like, GATOR1 complex subunit; NPR3-like; NPR3-like, GATOR1 complex subunit; NPRL3; Phg; Protein CGTHBA; Prox1; RMD11; RP23-177M10.6
Gene Symbols NPRL3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NPRL3. Recombinant protein control fragment (Product #RP-88735).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8131
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.