Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NPRL3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598424
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598424 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598424 Supplier Invitrogen™ Supplier No. PA598424
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NPRL3 Polyclonal Antibody for ELISA, IHC (P), Western Blot

The function of the encoded protein is not known.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NPRL3
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4
Gene NPRL3
Gene Accession No. Q12980
Gene Alias -14 gene protein; Aag; alpha-globin regulatory element-containing gene protein; alphaRE-14; C16orf35; CGTHBA; conserved gene telomeric to alpha globin cluster; GATOR complex protein NPRL3; HS-26; HS40; HS-40; MARE; nitrogen permease regulator 3-like protein; NPR3; NPR3 like, GATOR1 complex subunit; NPR3-like; NPR3-like, GATOR1 complex subunit; NPRL3; Phg; Protein CGTHBA; Prox1; RMD11; RP23-177M10.6
Gene Symbols NPRL3
Host Species Rabbit
Immunogen Recombinant Human GATOR complex protein NPRL3 protein (349-482AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8131
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.