Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NPRL3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA537905
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA537905 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA537905 Supplier Invitrogen™ Supplier No. PA537905
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is predicted to react with mouse and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 16,000.

The function of the encoded protein is not known.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NPRL3
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene NPRL3
Gene Accession No. Q12980
Gene Alias -14 gene protein; Aag; alpha-globin regulatory element-containing gene protein; alphaRE-14; C16orf35; CGTHBA; conserved gene telomeric to alpha globin cluster; GATOR complex protein NPRL3; HS-26; HS40; HS-40; MARE; nitrogen permease regulator 3-like protein; NPR3; NPR3 like, GATOR1 complex subunit; NPR3-like; NPR3-like, GATOR1 complex subunit; NPRL3; Phg; Protein CGTHBA; Prox1; RMD11; RP23-177M10.6
Gene Symbols NPRL3
Host Species Goat
Immunogen Peptide with sequence C-QRSQEHPASQTNKP, from the internal region near the N-terminus of mouse Nprl3 (aa 29-42).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8131
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.