Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva nProtein A Sepharose™ 4 Fast Flow Affinity Media
GSA_VA

Catalog No. 45002981 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
5 mL
pH Range:
3 to 9
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity pH Range
45-002-981 5 mL 3 to 9
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45-002-981 Supplier Cytiva Supplier No. 17528001
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Used for purification of monoclonal and polyclonal antibodies. Cytiva Lifescience™ nProtein A Sepharose™ 4 Fast Flow Affinity Media contains native protein A coupled to Sepharose 4 Fast Flow.

  • Low leakage of protein A with nearly twice total IgG binding capacity of Protein A Sepharose CL-4B
  • Used in large-scale FDA-approved processes
  • Manufactured without using animal-derived components
  • Rigidity and high degree of substitution of Sepharose 4 Fast Flow matrix facilitates speedy process of large volumes dilute cell culture fluid
  • High chemical, thermal and mechanical stability
  • Withstands high concentrations of hydrogen bond disrupting agents

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 5 mL
For Use With (Application) Purification of Monoclonal and Polyclonal Antibodies
Stability No significant change in chromatographic performance after 1 week storage using 8 M Urea, 6 M Gua-HCI, 2% Benzyl Alcohol or 20% Ethanol
Storage Requirements 2°C to 8°C
Binding Capacity >35 mg Human IgG/mL
Product Type nProtein A Sepharose 4 Fast Flow Media
Flow Rate 150 to 250 cm/Hoμr
Particle Size 90 μm
Height (English) Bed 9.84 in.
Height (Metric) Bed 25 cm
Max. Pressure 100 kPa
Matrix 4% Cross-Linked Agarose
pH Range 3 to 9
Storage Buffer 20% Ethanol
Ligand Type Native Protein A (S. aureus)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.