Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NPSR1/GPR154 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981360
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1360 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1360 Supplier Bioss Supplier No. BS11430R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

May be active in signaling pathway in an autocrine or paracrine fashion in several tissues. Receptor for neuropeptide S, it may mediate its action, such as inhibitory effects, on cell growth. Involved in pathogenesis of asthma and other IgE-mediated diseases.

Specifications

Antigen NPSR1/GPR154
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene NPSR1
Gene Accession No. Q6W5P4
Gene Alias 9330128H10Rik; ASRT2; G protein-coupled receptor 154; G protein-coupled receptor PGR14; GPR154; GPRA; G-protein coupled receptor 154; G-protein coupled receptor for asthma susceptibility; G-protein coupled receptor PGR14; MVTR; Neuropeptide S receptor; neuropeptide S receptor 1; NPSR; Npsr1; PGR14; RGD1564154; vasopressin receptor-related receptor 1; vasotocin receptor; VRR1
Gene Symbols NPSR1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human NPSR1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 387129
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.