Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NPSR1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA533986
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA533986 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA533986 Supplier Invitrogen™ Supplier No. PA533986
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey, Marmoset, Mouse, Rat, Hamster (100%); Elephant, Panda, Bovine, Bat, Pig, Turkey, Chicken (94%); Dog, Opossum, Lizard (89%); Horse, Platypus (83%). Proteinase K treatment is required for antigen retrieval.

May be active in signaling pathway in an autocrine or paracrine fashion in several tissues. Receptor for neuropeptide S, it may mediate its action, such as inhibitory effects, on cell growth. Involved in pathogenesis of asthma and other IgE-mediated diseases.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NPSR1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene NPSR1
Gene Accession No. P0C0L6, Q6W5P4, Q8BZP8
Gene Alias 9330128H10Rik; ASRT2; G protein-coupled receptor 154; G protein-coupled receptor PGR14; Gpr154; GPRA; G-protein coupled receptor 154; G-protein coupled receptor for asthma susceptibility; G-protein coupled receptor PGR14; MVTR; Neuropeptide S receptor; neuropeptide S receptor 1; NPSR; Npsr1; Pgr14; RGD1564154; vasopressin receptor-related receptor 1; vasotocin receptor; VRR1
Gene Symbols NPSR1
Host Species Rabbit
Immunogen Synthetic 18 amino acid peptide from 2nd extracellular domain of human NPSR1.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100765143, 300458, 319239, 387129
Target Species Human, Mouse, Rat, Monkey, Hamster
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.