Learn More
NPT1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
| Antigen | NPT1 |
|---|---|
| Concentration | 0.3 mg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-3064
![]()
|
Proteintech Group Inc
207511AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Western blot analysis using this antibody detected a major band around 45-55 kDa in kidney tissue.
The function of this protein remains unknown.Specifications
| NPT1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| Q14916, Q61983, Q62795 | |
| 171080, 20504, 6568 | |
| NPT1 Fusion Protein Ag12819 | |
| Primary | |
| -20°C | |
| SLC17A1 |
| 0.3 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Na(+)/PI cotransporter 1, Na/Pi 4, NAPI 1, SLC17A1 | |
| Slc17a1 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.