Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NPTN Monoclonal Antibody (OTI2F4), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA504435S


Description
NPTN Monoclonal antibody specifically detects NPTN in Human samples. It is validated for Immunocytochemistry, Western BlotSpecifications
NPTN | |
Monoclonal | |
0.81 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
P36955 | |
Mouse | |
Affinity chromatography | |
RUO | |
5176 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG1 |
Immunocytochemistry, Western Blot | |
OTI2F4 | |
Unconjugated | |
SERPINF1 | |
AW554172; glycoprotein 55; glycoprotein 55/65; glycoprotein 65; GP55; gp55/65; GP65; Neuroplastin; np55; np65; NPTN; Sdfr1; Sdr1; SDR-1; stromal cell derived factor receptor 1; stromal cell-derived receptor 1 | |
Full length human recombinant protein of human SERPINF1 produced in HEK293T cell. | |
30 μL | |
Primary | |
Human | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction