Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NPTX1 Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA5143091
This item is not returnable.
View return policy
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:128,000.
NPTX1 is a member of the neuronal pentraxin family. Human neuronal pentraxin 1 is similar to rat NP1 which is a binding protein for the snake venom toxin taipoxin.
Specifications
| NPTX1 | |
| Polyclonal | |
| Unconjugated | |
| Nptx1 | |
| 47 kDa taipoxin-binding protein; D11Bwg1004e; neuronal pentraxin 1; neuronal pentraxin I; Neuronal pentraxin-1; NP1; NP-I; NPTX1 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 4884 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q15818 | |
| Nptx1 | |
| Peptide with sequence C-ERVKIETALTSLHQR (aa 197-211). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction