Learn More
Invitrogen™ NPTX1 Polyclonal Antibody

Description
NPTX1 Polyclonal Antibody for IHC (P), Western Blot
Specifications
Specifications
| Antigen | NPTX1 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.33 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | Nptx1 |
| Gene Accession No. | P47971, Q15818, Q62443 |
| Gene Alias | 47 kDa taipoxin-binding protein; D11Bwg1004e; neuronal pentraxin 1; neuronal pentraxin I; Neuronal pentraxin-1; NP1; NP-I; NPTX1 |
| Gene Symbols | Nptx1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.