Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NPTX1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA585018
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA585018 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA585018 Supplier Invitrogen™ Supplier No. PA585018
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ETLSQLGQTL QSLKTRLENL EQYSRLNSSS QTNSLKDLLQ SKIDELERQV LSRVNT Highest antigen sequence indentity to the following orthologs: Rat - 98%, Mouse - 98%.

NPTX1 is a member of the neuronal pentraxin family. Human neuronal pentraxin 1 is similar to rat NP1 which is a binding protein for the snake venom toxin taipoxin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NPTX1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Nptx1
Gene Accession No. Q15818
Gene Alias 47 kDa taipoxin-binding protein; D11Bwg1004e; neuronal pentraxin 1; neuronal pentraxin I; Neuronal pentraxin-1; NP1; NP-I; NPTX1
Gene Symbols Nptx1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NPTX1. Recombinant protein control fragment (Product #RP-108286).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4884
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.