Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NPW Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 1 publication
$377.03 - $728.30
Specifications
| Antigen | NPW |
|---|---|
| Dilution | Immunohistochemistry-Paraffin 1:1000 - 1:2500 |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
NPW Polyclonal specifically detects NPW in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| NPW | |
| Polyclonal | |
| Rabbit | |
| Human | |
| HPPL8, L8, L8C, Neuropeptide W, PPL8, PPNPW, Prepro-Neuropeptide W, Preproprotein L8 | |
| NPW | |
| IgG | |
| Affinity Purified |
| Immunohistochemistry-Paraffin 1:1000 - 1:2500 | |
| Unconjugated | |
| RUO | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| 283869 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:REAPLLLPSWVQELWETRRRSSQAGIPVRAPRSPRAPEPALEPESLDFSGAGQRLRRDVSRPAVDPAANR | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title