Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NPW Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5115665
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5115665 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5115665 Supplier Thermo Scientific Supplier No. PA5115665
Add to Cart
Add to Cart

NPW Polyclonal Antibody

NPW Polyclonal Antibody for Western Blot, ICC/IF

TRUSTED_SUSTAINABILITY

Specifications

Antigen NPW
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene NPW
Gene Accession No. Q8N729
Gene Alias Gm935; hPPL8; L8; L8C; Neuropeptide W; Neuropeptide W-23; Neuropeptide W-30; NPW; NPW23; NPW30; PP PPNPW; Ppl8; PPNPW; prepro-neuropeptide W; prepro-Neuropeptide W polypeptide; preproprotein L8
Gene Symbols NPW
Host Species Rabbit
Immunogen A synthesized peptide derived from human NPW(Accession Q8N729), corresponding to amino acid residues Q96-P146.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 283869
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.