Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NPY1R Antibody (556153), DyLight 550, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB6400L
Description
NPY1R Monoclonal antibody specifically detects NPY1R in Human samples. It is validated for Flow Cytometry, CyTOF-reportedSpecifications
NPY1R | |
Monoclonal | |
DyLight 550 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry, CyTOF | |
556153 | |
Flow Cytometry, CyTOF-reported | |
neuropeptide Y receptor type 1, neuropeptide Y receptor Y1, NPY1-R, NPYR, NPYY1 | |
NS0 mouse myeloma cell line transfected with human NPY1R, Met1-Ile384, Accession # P25929 | |
0.1 mL | |
GPCR, Neuroscience, Neurotransmission | |
4886 | |
Store at 4°C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction