Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NPY1R Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110989
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110989 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110989 Supplier Invitrogen™ Supplier No. PA5110989
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KRRNNMMDKM RDNKYRSSET KRINI Highest antigen sequence indentity to the following orthologs: Mouse - 92%, Rat - 88%.

Neuropeptide Y receptors are a class of G-protein coupled receptors which are activated by the closely related peptide horomones neuropeptide Y, peptide YY and pancreatic polypeptide. These receptors are involved in control of a diverse set of behavioral processes including appetite, circadian rhythme, and anxiety.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NPY1R
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NPY1R
Gene Accession No. P25929
Gene Alias FC5; Neuropepetide Y1 Receptor; neuropeptide Y receptor subtype; neuropeptide Y receptor type 1; neuropeptide Y receptor Y1; neuropeptide Y1 receptor; NPY-1; NPY-1 receptor; Npy1r; NPY1-R; NPYR; NPYY1; NPY-Y1 receptor; PY receptor Y1; Y1-R
Gene Symbols NPY1R
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human NPY1R. Recombinant protein control fragment (Product #RP-95314).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4886
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.