Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NPY2R Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA595856
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA595856 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA595856 Supplier Invitrogen™ Supplier No. PA595856
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LKEYKLIFTV FHIIAMCSTF ANPLLYGWMN SNYRKAFLSA FRCEQRLDAI HSEVSVTFKA KKNLEVRKNS GPNDSFTEAT NV.

Receptor for neuropeptide Y and peptide YY. The rank order of affinity of this receptor for pancreatic polypeptides is PYY > NPY > PYY (3-36) > NPY (2-36) > [Ile-31, Gln-34] PP > [Leu-31, Pro-34] NPY > PP, [Pro-34] PYY and NPY free acid.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NPY2R
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.23 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene NPY2R
Gene Accession No. P49146
Gene Alias MGC124241; MGC124242; MGC124244; Neuropeptide Y receptor type 2; neuropeptide Y receptor Y2; neuropeptide Y receptor Y2-like; neuropeptide Y/peptide YY-Y2 receptor; Neuropeptide y-y2; neuropeptide Y-Y2 receptor; NPY2R; NPY2-R; NPY-Y2 receptor; Y2 receptor
Gene Symbols NPY2R
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 300 to the C-terminus of human NPY2R (NP_0009011).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4887
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.