Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NPY2R Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598014
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598014 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598014 Supplier Invitrogen™ Supplier No. PA598014
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NPY2R Polyclonal Antibody for Western Blot, ICC/IF, ELISA

Receptor for neuropeptide Y and peptide YY. The rank order of affinity of this receptor for pancreatic polypeptides is PYY > NPY > PYY (3-36) > NPY (2-36) > [Ile-31, Gln-34] PP > [Leu-31, Pro-34] NPY > PP, [Pro-34] PYY and NPY free acid.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NPY2R
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 4.73 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene NPY2R
Gene Accession No. P49146
Gene Alias MGC124241; MGC124242; MGC124244; Neuropeptide Y receptor type 2; neuropeptide Y receptor Y2; neuropeptide Y receptor Y2-like; neuropeptide Y/peptide YY-Y2 receptor; Neuropeptide y-y2; neuropeptide Y-Y2 receptor; NPY2R; NPY2-R; NPY-Y2 receptor; Y2 receptor
Gene Symbols NPY2R
Host Species Rabbit
Immunogen Recombinant Human Neuropeptide Y receptor type 2 protein (326-381AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4887
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.