Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NPY4R Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA534106
Description
Percent identity with other species by BLAST analysis: Human, Gibbon (100%) Gorilla, Panda (94%) Monkey, Bovine, Horse, Pig (88%) Bat, Rabbit (82%).
Receptor for neuropeptide Y and peptide YY. The rank order of affinity of this receptor for pancreatic polypeptides is PP, PP (2-36) and [Ile-31, Gln-34] PP > [Pro-34] PYY > PYY and [Leu-31, Pro-34] NPY > NPY > PYY (3-36) and NPY (2-36) > PP (13- 36) > PP (31-36) > NPY free acid.
Specifications
NPY4R | |
Polyclonal | |
Unconjugated | |
NPY4R | |
neuropeptide Y receptor type 4; neuropeptide Y receptor Y4; NPY4R; NPY4-R; NPYR-D; NYYR-D; Pancreatic polypeptide receptor 1; PP1; PPYR1; Y4; Y4 receptor | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
5540 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
P50391 | |
NPY4R | |
Synthetic 17 amino acid peptide from C-terminus of human PPYR1. | |
50 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction