Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NPY4R Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596949
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596949 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596949 Supplier Invitrogen™ Supplier No. PA596949
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MNTSHLLALL LPKSPQGENR SKPLGTPYNF SEHCQDSVDV MVFIVTSYSI ETVVGVLGNL CLMCVTVRQK EKANVTNLLI; Positive Samples: MCF-7, SH-SY5Y, SW480, HT-29; Cellular Location: Cell membrane, Multi-pass membrane protein.

Receptor for neuropeptide Y and peptide YY. The rank order of affinity of this receptor for pancreatic polypeptides is PP, PP (2-36) and [Ile-31, Gln-34] PP > [Pro-34] PYY > PYY and [Leu-31, Pro-34] NPY > NPY > PYY (3-36) and NPY (2-36) > PP (13- 36) > PP (31-36) > NPY free acid.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NPY4R
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.75 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene NPY4R
Gene Accession No. P50391
Gene Alias neuropeptide Y receptor type 4; neuropeptide Y receptor Y4; NPY4R; NPY4-R; NPYR-D; NYYR-D; Pancreatic polypeptide receptor 1; PP1; PPYR1; Y4; Y4 receptor
Gene Symbols NPY4R
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-80 of human NPY4R (NP_0059634).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5540
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.