Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NPY5R Antibody, Novus Biologicals™
SDP

Catalog No. NB1001538 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1001538 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1001538 Supplier Novus Biologicals Supplier No. NB1001538
Only null left
Add to Cart
Add to Cart

Goat Polyclonal Antibody has been used in 5 publications

NPY5R Polyclonal specifically detects NPY5R in Human, Rat, Hamster samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Peptide ELISA.

Specifications

Antigen NPY5R
Applications Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.3 - 1 μg/mL, Immunohistochemistry 4 - 6 μg/mL, Immunoprecipitation, Immunohistochemistry-Paraffin 4 - 6 μg/mL, Peptide ELISA Detection limit 1:32000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. Q15761
Gene Alias neuropeptide Y receptor type 5, neuropeptide Y receptor Y5, NPYR5NPY5-R, NPY-Y5 receptor, NPYY5-R, Y5 receptor
Gene Symbols NPY5R
Host Species Goat
Immunogen Peptide with sequence C-GLSNKENRLEEN corresponding to internal region according to NP_006165.1.
Molecular Weight of Antigen 51 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline GPCR
Primary or Secondary Primary
Gene ID (Entrez) 4889
Test Specificity Human and Rat. Expected from sequence similarity: Mouse, Dog.
Target Species Human, Rat, Hamster
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.