Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NPY5R Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA533992
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA533992 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA533992 Supplier Invitrogen™ Supplier No. PA533992
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey (100%) Marmoset, Bovine, Panda (88%) Dog, Rabbit, Pig (82%).

Receptor for neuropeptide Y and peptide YY. The activity of this receptor is mediated by G proteins that inhibit adenylate cyclase activity. Seems to be associated with food intake. Could be involved in feeding disorders.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NPY5R
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene Npy5r
Gene Accession No. Q15761
Gene Alias HGNC:7958; Neuropeptide Y receptor type 5; neuropeptide Y receptor type 5-like protein; neuropeptide Y receptor Y5; Neuropeptide Y5 receptor; Npy5; NPY5 receptor; Npy5r; NPY5-R; NPYR5; NPYY5; NPY-Y5 receptor; NPYY5-R; Y5 receptor; Y5R
Gene Symbols Npy5r
Host Species Rabbit
Immunogen Synthetic 17 amino acid peptide from 3rd cytoplasmic domain of human NPY5R.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4889
Target Species Human, Monkey
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.