Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NPY5R Polyclonal Antibody, Biotin
GREENER_CHOICE

Catalog No. PIPA5142150
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5142150 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5142150 Supplier Invitrogen™ Supplier No. PA5142150
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:32,000.

Receptor for neuropeptide Y and peptide YY. The activity of this receptor is mediated by G proteins that inhibit adenylate cyclase activity. Seems to be associated with food intake. Could be involved in feeding disorders.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NPY5R
Applications Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Biotin
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene Npy5r
Gene Accession No. Q15761
Gene Alias HGNC:7958; Neuropeptide Y receptor type 5; neuropeptide Y receptor type 5-like protein; neuropeptide Y receptor Y5; Neuropeptide Y5 receptor; Npy5; NPY5 receptor; Npy5r; NPY5-R; NPYR5; NPYY5; NPY-Y5 receptor; NPYY5-R; Y5 receptor; Y5R
Gene Symbols Npy5r
Host Species Goat
Immunogen Peptide with sequence C-GLSNKENRLEEN (aa 241-252).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4889
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.