Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NQO-2 Antibody (1E3), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP174033B
Description
NQO-2 Monoclonal specifically detects NQO-2 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
NQO-2 | |
Monoclonal | |
Biotin | |
PBS with 0.05% Sodium Azide | |
NQO2 | |
Recombinant human NQO2 (1-231aa) purified from E. coli | |
100 μL | |
Stem Cell Markers | |
4835 | |
Human, Mouse | |
Purified |
Western Blot, ELISA | |
1000 | |
Western Blot, ELISA | |
dioxin-inducible-2, EC 1.10.99.2, NAD(P)H dehydrogenase, quinone 2, NAD(P)H menadione oxidoreductase 2, dioxin-inducible, NRH dehydrogenase [quinone] 2, NRH:quinone oxidoreductase 2, Quinone reductase 2, ribosyldihydronicotinamide dehydrogenase, ribosyldihydronicotinamide dehydrogenase [quinone] | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Anti-human NQO2 mAb, clone 1E3, is derived from hybridization of mouse FO myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human NQO2 protein. | |
Store at 4C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction