Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NQO-2 Antibody (OTI3C11), Alexa Fluor™ 532, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273045AF532
Description
NQO-2 Monoclonal specifically detects NQO-2 in Human, Canine, Monkey samples. It is validated for Western Blot.Specifications
NQO-2 | |
Monoclonal | |
Alexa Fluor 532 | |
50 mM sodium borate with 0.05% sodium azide | |
NQO2 | |
Full length recombinant protein of human NQO-2 (NP_000895) produced in HEK293T cells. | |
Primary | |
Human, Canine, Monkey | |
IgG1 |
Western Blot | |
OTI3C11 | |
Western Blot | |
dioxin-inducible-2, EC 1.10.99.2, NAD(P)H dehydrogenase, quinone 2, NAD(P)H menadione oxidoreductase 2, dioxin-inducible, NRH dehydrogenase [quinone] 2, NRH:quinone oxidoreductase 2, Quinone reductase 2, ribosyldihydronicotinamide dehydrogenase, ribosyldihydronicotinamide dehydrogenase [quinone] | |
Mouse | |
0.1 mL | |
4835 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction