Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NQO-2 Antibody (OTI3G4), Alexa Fluor™ 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273049AF350
Description
NQO-2 Monoclonal specifically detects NQO-2 in Human, Mouse, Canine, Primate, Monkey samples. It is validated for Western Blot, Immunohistochemistry.Specifications
| NQO-2 | |
| Monoclonal | |
| Alexa Fluor 350 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| NQO2 | |
| Full length recombinant protein of human NQO-2 (NP_000895) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Canine, Primate, Monkey | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI3G4 | |
| Western Blot, Immunohistochemistry | |
| dioxin-inducible-2, EC 1.10.99.2, NAD(P)H dehydrogenase, quinone 2, NAD(P)H menadione oxidoreductase 2, dioxin-inducible, NRH dehydrogenase [quinone] 2, NRH:quinone oxidoreductase 2, Quinone reductase 2, ribosyldihydronicotinamide dehydrogenase, ribosyldihydronicotinamide dehydrogenase [quinone] | |
| Mouse | |
| Affinity Purified | |
| Stem Cell Markers | |
| 4835.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction