Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NQO1 Mouse anti-Human, Unconjugated, Clone: A180, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MABS2176100UG
Description
NQO1 Monoclonal antibody specifically detects NQO1 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blotting.
Specifications
NQO1 | |
Monoclonal | |
Unconjugated | |
NAD(P)H dehydrogenase [quinone] 1;EC:1.6.5.2;Azoreductase;DT-diaphorase;DTD;Menadione reductase;NAD(P)H:quinone oxidoreductase 1;Phylloquinone reductase;Quinone reductase 1;QR1 | |
Mouse | |
∼31 kDa observed; 30.87 kDa calculated. Uncharacterized bands may be observed in some lysate(s). | |
100 μg | |
Primary | |
Human | |
Purified |
Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
A180 | |
NP_000894 | |
NQO1, DIA4, NMOR1 | |
Full-length recombinant human NAD(P)H dehydrogenase (quinone) 1 (NQO1). | |
Protein G Purified | |
Signaling | |
Clone A180 is a mouse monoclonal antibody that specifically detects human NAD(P)H dehydrogenase [quinone] 1 (NQO1). | |
Stable for 1 year at 2 to 8°C from date of receipt. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction