Learn More
Invitrogen™ NQO1 Recombinant Rabbit Monoclonal Antibody (7L9B4)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA551434
Description
Immunogen sequence: MVGRRALIVL AHSERTSFNY AMKEAAAAAL KKKGWEVVES DLYAMNFNPI ISRKDITGKL KDPANFQYPA ESVLAYKEGH LSPDIVAEQK KLEAADLVIF QFPLQWFGVP AILKGWFERV FIGEFAYTYA AMYDKGPFRS KKAVLSITTG GSGSMYSLQG IHGDMNVILW PIQSGILHFC GFQVLEPQLT YSIGHTPADA RIQILEGWKK RLENIWDETP LYFAPSSLFD LNFQAGFLMK KEVQDEEKNK KFGLSVGHHL GKSIPTDNQI KARK.
NQO1 is a member of the NAD(P)H dehydrogenase (quinone) family and a cytoplasmic 2-electron reductase. This FAD-binding protein forms homodimers and reduces quinones to hydroquinones. NQO1's enzymatic activity prevents the one electron reduction of quinones that results in the production of radical species. Altered expression of the protein has been seen in many tumors and is also associated with Alzheimer's disease (AD).
Specifications
NQO1 | |
Recombinant Monoclonal | |
0.24 mg/mL | |
PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
P05982, P15559, Q64669 | |
NQO1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-274 of human NQO1 (NP_000894.1). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
Liquid |
ELISA, Western Blot, Immunocytochemistry | |
7L9B4 | |
Unconjugated | |
NQO1 | |
AV001255; Azoreductase; DHQU; DIA4; Diaphorase (NADH/NADPH); diaphorase (NADH/NADPH) (cytochrome b-5 reductase); Diaphorase 4; diaphorase 4 (NADH/NADPH); diaphorase-4; dioxin-inducible 1; DTD; DT-diaphorase; menadione reductase; NAD(P)H dehydrogenase (quinone); NAD(P)H dehydrogenase [quinone] 1; NAD(P)H dehydrogenase, quinone 1; NAD(P)H menadione oxidoreductase 1, dioxin inducible; NAD(P)H quinone dehydrogenase 1; NAD(P)H: quinone oxidoreductase 1; NAD(P)H:menadione oxidoreductase; NAD(P)H:menadione oxidoreductase 1; NAD(P)H:Quinone acceptor oxidoreductase type 1; NAD(P)H:quinone oxidoreductase 1; NAD(P)H:quinone oxireductase; NADPH-d; nicotinamide adenine dinucleotide phosphate diaphorase; Nmo1; Nmo-1; Nmor1; NMORI; NQ01; NQO1; Ox1; Ox-1; phylloquinone reductase; QR1; quinone reductase 1 | |
Rabbit | |
Affinity chromatography | |
RUO | |
1728, 18104, 24314 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.