Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NQO1 Recombinant Rabbit Monoclonal Antibody (JF440-1)
GREENER_CHOICE

Catalog No. PIMA532528
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA532528 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA532528 Supplier Invitrogen™ Supplier No. MA532528
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.

NQO1 is a member of the NAD(P)H dehydrogenase (quinone) family and a cytoplasmic 2-electron reductase. This FAD-binding protein forms homodimers and reduces quinones to hydroquinones. NQO1's enzymatic activity prevents the one electron reduction of quinones that results in the production of radical species. Altered expression of the protein has been seen in many tumors and is also associated with Alzheimer's disease (AD).
TRUSTED_SUSTAINABILITY

Specifications

Antigen NQO1
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Western Blot
Classification Recombinant Monoclonal
Clone JF440-1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.05% BSA, 40% Glycerol and 0.05% sodium azide; pH 7.4
Gene NQO1
Gene Accession No. P05982, P15559, Q64669
Gene Alias AV001255; Azoreductase; DHQU; DIA4; Diaphorase (NADH/NADPH); diaphorase (NADH/NADPH) (cytochrome b-5 reductase); Diaphorase 4; diaphorase 4 (NADH/NADPH); diaphorase-4; dioxin-inducible 1; DTD; DT-diaphorase; menadione reductase; NAD(P)H dehydrogenase (quinone); NAD(P)H dehydrogenase [quinone] 1; NAD(P)H dehydrogenase, quinone 1; NAD(P)H menadione oxidoreductase 1, dioxin inducible; NAD(P)H quinone dehydrogenase 1; NAD(P)H: quinone oxidoreductase 1; NAD(P)H:menadione oxidoreductase; NAD(P)H:menadione oxidoreductase 1; NAD(P)H:Quinone acceptor oxidoreductase type 1; NAD(P)H:quinone oxidoreductase 1; NAD(P)H:quinone oxireductase; NADPH-d; nicotinamide adenine dinucleotide phosphate diaphorase; Nmo1; Nmo-1; Nmor1; NMORI; NQ01; NQO1; Ox1; Ox-1; phylloquinone reductase; QR1; quinone reductase 1
Gene Symbols NQO1
Host Species Rabbit
Immunogen Synthetic peptide within Human NQO1 aa 1-49.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1728, 18104, 24314
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.