Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NQO2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554387
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554387 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554387 Supplier Invitrogen™ Supplier No. PA554387
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MAGKKVLIVY AHQEPKSFNG SLKNVAVDEL SRQGCTVTVS DLYAMNLEPR ATDKDITGTL SNPEVFNYGV ETHEAYKQRS LASDITDEQK KVREADLVIF QFPLYWFSVP A Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 79%.

NQO2 is a flavoprotein that catalyzes the 2-electron reduction of various quinones, redox dyes, and the vitamin K menadione. NQO2 predominantly uses dihydronicotinamide riboside (NRH) as the electron donor (Wu et al., 1997).
TRUSTED_SUSTAINABILITY

Specifications

Antigen NQO2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NQO2
Gene Accession No. P16083
Gene Alias DHQV; DIA6; EC 1.10.99.2; MGC94180; NAD(P)H dehydrogenase, quinone 2; NAD(P)H menadione oxidoreductase 2, dioxin inducible; NAD(P)H menadione oxidoreductase-1, dioxin-inducible-2; NAD(P)H quinone dehydrogenase 2; NMO2; NMOR2; NQO 2; Nqo2; NRH dehydrogenase; NRH dehydrogenase [quinone] 2; NRH: quinone oxidoreductase; NRH:quinone oxidoreductase 2; N-ribosyldihydronicotinamide:quinone reductase 2; OTTHUMP00000015948; OTTHUMP00000015949; OTTHUMP00000015953; Ox 2; Ox2; QR2; Quinone; Quinone reductase 2; quinone ribosyldihydronicotinamide dehydrogenase; ribosyldihydronicotinamide; ribosyldihydronicotinamide dehydrogenase; ribosyldihydronicotinamide dehydrogenase [quinone]; ribosyldihydronicotinamide dehydrogenase quinone
Gene Symbols NQO2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NQO2. Recombinant protein control fragment (Product #RP-93141).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4835
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.