Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NR0B1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA595912
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA595912 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA595912 Supplier Invitrogen™ Supplier No. PA595912
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MAGENHQWQG SILYNMLMSA KQTRAAPEAP ETRLVDQCWG CSCGDEPGVG REGLLGGRNV ALLYRCCFCG KDHPRQGSIL YSMLTSAKQT YAAPKAPEAT LGPCWGCSCG SDPGVGRAGL PGGRPVALLY RCCFCGEDHP RQGSILYSLL TSSKQTHVAP AAPEARPGGA WWDRSYFAQR PGGKEALPGG RATALLYRCC FCGEDHPQQG STLYCVPTST NQAQAAPEER PRAPWWDTSS GALRPVALKS PQVVCEAASA; Positive Samples: HeLa, Jurkat, MCF7, HepG2, Raji, Mouse spleen; Cellular Location: Cytoplasm, Nucleus.

This gene encodes a protein that contains a DNA-binding domain. The encoded protein acts as a dominant-negative regulator of transcription which is mediated by the retinoic acid receptor. This protein also functions as an anti-testis gene by acting antagonistically to Sry. Mutations in this gene result in both X-linked congenital adrenal hypoplasia and hypogonadotropic hypogonadism.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NR0B1
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 2.96 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene NR0B1
Gene Accession No. P51843, Q61066
Gene Alias adrenal hypoplasia, congenital homolog; AHC; Ahch; AHX; DAX; DAX 1; Dax1; DAX-1; DSS; DSS-AHC critical region on the X chromosome protein 1; GTD; HHG; Nr0b1; NROB1; nuclear hormone receptor; nuclear receptor DAX-1; Nuclear receptor subfamily 0 group B member 1; nuclear receptor subfamily 0, group B, member 1; orphan nuclear receptor DAX-1; R0B1; SRXY2
Gene Symbols NR0B1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-260 of human NR0B1 (NP_0004662).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11614, 190
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.