Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NR0B1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA533466
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA533466 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA533466 Supplier Invitrogen™ Supplier No. PA533466
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human, Chimpanzee, Orangutan, Monkey (100%) Gorilla (95%) Gibbon (90%) Marmoset (85%).

This gene encodes a protein that contains a DNA-binding domain. The encoded protein acts as a dominant-negative regulator of transcription which is mediated by the retinoic acid receptor. This protein also functions as an anti-testis gene by acting antagonistically to Sry. Mutations in this gene result in both X-linked congenital adrenal hypoplasia and hypogonadotropic hypogonadism.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NR0B1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene NR0B1
Gene Accession No. P51843
Gene Alias adrenal hypoplasia, congenital homolog; AHC; Ahch; AHX; DAX; DAX 1; Dax1; DAX-1; DSS; DSS-AHC critical region on the X chromosome protein 1; GTD; HHG; Nr0b1; NROB1; nuclear hormone receptor; nuclear receptor DAX-1; Nuclear receptor subfamily 0 group B member 1; nuclear receptor subfamily 0, group B, member 1; orphan nuclear receptor DAX-1; R0B1; SRXY2
Gene Symbols NR0B1
Host Species Rabbit
Immunogen Synthetic 20 amino acid peptide from C-terminus of human DAX1.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 190
Target Species Human, Monkey
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.