Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NR0B1 Recombinant Rabbit Monoclonal Antibody (JU33-05)
GREENER_CHOICE

Catalog No. PIMA534635
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA534635 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA534635 Supplier Invitrogen™ Supplier No. MA534635
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Positive Control: A549, Hela, SH-SY-5Y.

This gene encodes a protein that contains a DNA-binding domain. The encoded protein acts as a dominant-negative regulator of transcription which is mediated by the retinoic acid receptor. This protein also functions as an anti-testis gene by acting antagonistically to Sry. Mutations in this gene result in both X-linked congenital adrenal hypoplasia and hypogonadotropic hypogonadism.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NR0B1
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone JU33-05
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 40% glycerol, 0.05% BSA and 0.05% sodium azide; pH 7.4
Gene NR0B1
Gene Accession No. P51843
Gene Alias adrenal hypoplasia, congenital homolog; AHC; Ahch; AHX; DAX; DAX 1; Dax1; DAX-1; DSS; DSS-AHC critical region on the X chromosome protein 1; GTD; HHG; Nr0b1; NROB1; nuclear hormone receptor; nuclear receptor DAX-1; Nuclear receptor subfamily 0 group B member 1; nuclear receptor subfamily 0, group B, member 1; orphan nuclear receptor DAX-1; R0B1; SRXY2
Gene Symbols NR0B1
Host Species Rabbit
Immunogen Synthetic peptide within Human NR0B1 aa 1-50.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 190
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles, store in dark
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.