Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NR0B2 Monoclonal Antibody (OTI7B5)
GREENER_CHOICE

Catalog No. PIMA526786
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA526786 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA526786 Supplier Invitrogen™ Supplier No. MA526786
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NR0B2 Monoclonal Antibody for Western Blot, IHC (P)

The protein encoded by this gene is an unusual orphan receptor that contains a putative ligand-binding domain but lacks a conventional DNA-binding domain. The gene product is a member of the nuclear hormone receptor family, a group of transcription factors regulated by small hydrophobic hormones, a subset of which do not have known ligands and are referred to as orphan nuclear receptors. The protein has been shown to interact with retinoid and thyroid hormone receptors, inhibiting their ligand-dependent transcriptional activation. In addition, interaction with estrogen receptors has been demonstrated, leading to inhibition of function. Studies suggest that the protein represses nuclear hormone receptor-mediated transactivation via two separate steps: competition with coactivators and the direct effects of its transcriptional repressor function.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NR0B2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI7B5
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene NR0B2
Gene Accession No. Q15466
Gene Alias Nr0b2; Nr0b2a; nuclear receptor SHP; nuclear receptor subfamily 0 group B member 2; nuclear receptor subfamily 0, group B, member 2; orphan nuclear receptor SHP; SHP; SHP1; SHP-1; small heterodimer partner; small heterodimer partner homologue
Gene Symbols NR0B2
Host Species Mouse
Immunogen Full length human recombinant protein of NR0B2 produced in E.coli.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8431
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.