Learn More
Invitrogen™ NR1D1 Monoclonal Antibody (2G6)

Description
Peptide Sequence: SQCPLETSPT QHPTPGPMGP SPPPAPVPSP LVGFSQFPQQ LTPPRSPSPE PTVEDVISQV ARAHREIFTY AHDKLGSSPG NFNANHASG.
Specifications
Specifications
| Antigen | NR1D1 |
| Applications | ELISA, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 2G6 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | NR1D1 |
| Gene Accession No. | P20393 |
| Gene Alias | A530070C09Rik; BD73; Ear1; ear-1; EAR-1R; HREV; NR1D1; nuclear receptor protein; nuclear receptor Rev-ErbA-alpha; nuclear receptor subfamily 1 group D member 1; nuclear receptor subfamily 1, group D, member 1; orphan nuclear receptor; R75201; rev-erb alpha; Rev-erba; rev-erbA alpha; rev-erbA(alpha); Reverbaalpha; Rev-ErbAalpha; rev-erbA-alpha; Rev-ErbA-alpha protein; REV-ERBalpha; RVR; THRA1; THRAL; V-erbA-related protein 1; V-erbA-related protein EAR-1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.