Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NR1D1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA528737
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA528737 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA528737 Supplier Invitrogen™ Supplier No. PA528737
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: A431, H1299, HeLa, HepG2. Predicted reactivity: Mouse (100%), Rat (100%), Sheep (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes a member of the nuclear hormone receptor family, specifically the NR1 subfamily of receptors. The encoded protein functions as a transcriptional repressor and may play a role in circadian rhythms and carbohydrate and lipid metabolism. Alternatively spliced transcript variants have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NR1D1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.83 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene NR1D1
Gene Accession No. P20393
Gene Alias A530070C09Rik; BD73; Ear1; ear-1; EAR-1R; HREV; NR1D1; nuclear receptor protein; nuclear receptor Rev-ErbA-alpha; nuclear receptor subfamily 1 group D member 1; nuclear receptor subfamily 1, group D, member 1; orphan nuclear receptor; R75201; rev-erb alpha; Rev-erba; rev-erbA alpha; rev-erbA(alpha); Reverbaalpha; Rev-ErbAalpha; rev-erbA-alpha; Rev-ErbA-alpha protein; REV-ERBalpha; RVR; THRA1; THRAL; V-erbA-related protein 1; V-erbA-related protein EAR-1
Gene Symbols NR1D1
Host Species Rabbit
Immunogen Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of human NR1D1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9572
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.